siRNA Details
| Gene Symbol: Zfp950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029211.1 | - | 7 | 830 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp950| Gene Symbol: Zfp950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029211.1 | - | 7 | 830 | ||