siRNA Details
| Gene Symbol: Gm14005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028589.1 | 1 | 196 | |||
| NR_028590.1 | 1 | 196 | ||||
| NR_028591.1 | 1 | 196 | ||||
| GenBank mRNA | AK157218.1 | - | 1 | 196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14005| Gene Symbol: Gm14005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028589.1 | 1 | 196 | |||
| NR_028590.1 | 1 | 196 | ||||
| NR_028591.1 | 1 | 196 | ||||
| GenBank mRNA | AK157218.1 | - | 1 | 196 | ||