siRNA Details
| Gene Symbol: Spata5l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048180.1 | - | 1105 | |||
| AK133972.1 | - | 1088 | ||||
| AK169460.1 | - | 1125 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata5l1| Gene Symbol: Spata5l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048180.1 | - | 1105 | |||
| AK133972.1 | - | 1088 | ||||
| AK169460.1 | - | 1125 | ||||