siRNA Details
| Gene Symbol: Gm10804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040533.1 | 3 | 849 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10804| Gene Symbol: Gm10804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040533.1 | 3 | 849 | |||