siRNA Details
| Gene Symbol: 4631405J19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028444.1 | - | 1113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4631405J19Rik| Gene Symbol: 4631405J19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028444.1 | - | 1113 | |||