siRNA Details
Gene Symbol: 4930431P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045059.1 | 6 | 669 | |||
GenBank mRNA | AK015279.1 | - | 6 | 670 | ||
AK133263.1 | - | 4 | 2942 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930431P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045059.1 | 6 | 669 | |||
GenBank mRNA | AK015279.1 | - | 6 | 670 | ||
AK133263.1 | - | 4 | 2942 |