siRNA Details
| Gene Symbol: Haglr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110445.1 | 2 | 218 | |||
| GenBank mRNA | W45744.1 | - | 2 | 215 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI427313.1 | - | 218 | Chr. - - [-] on Build | ||

Haglr
Genomic Map