siRNA Details
| Gene Symbol: Gm13262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085384.1 | - | 2 | 1259 | ||
| AK143820.1 | - | 2 | 1287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13262| Gene Symbol: Gm13262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085384.1 | - | 2 | 1259 | ||
| AK143820.1 | - | 2 | 1287 | |||