siRNA Details
| Gene Symbol: Gm11529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033524.1 | 3 | 557 | |||
| GenBank mRNA | AK138406.1 | - | 3 | 558 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY724101.1 | - | 3 | 558 | Chr. 11 - 96448323 - 96464547 [-] on Build GRCm38 | |

Gm11529
Genomic Map