siRNA Details
| Gene Symbol: 4930456L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045887.1 | 2, 3 | 841 | |||
| GenBank mRNA | AK015480.1 | - | 2, 3 | 842 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930456L15Rik| Gene Symbol: 4930456L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045887.1 | 2, 3 | 841 | |||
| GenBank mRNA | AK015480.1 | - | 2, 3 | 842 | ||