siRNA Details
| Gene Symbol: Gm15498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC028660.1 | - | 2 | 694 | 1265093 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15498| Gene Symbol: Gm15498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC028660.1 | - | 2 | 694 | 1265093 | |