siRNA Details
| Gene Symbol: Rab10os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015551.1 | 6 | 528 | |||
| GenBank mRNA | AK005738.1 | - | 6 | 481 | ||
| AK007702.1 | - | 6 | 529 | |||
| BC080728.1 | - | 6 | 2325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab10os| Gene Symbol: Rab10os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015551.1 | 6 | 528 | |||
| GenBank mRNA | AK005738.1 | - | 6 | 481 | ||
| AK007702.1 | - | 6 | 529 | |||
| BC080728.1 | - | 6 | 2325 | |||