siRNA Details
| Gene Symbol: 2610203C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015470.1 | 2 | 72 | |||
| GenBank mRNA | AK009214.1 | - | 2 | 148 | ||
| AK011866.1 | - | 2 | 118 | |||
| AK082741.1 | - | 2 | 125 | |||
| BC037765.1 | - | 2 | 72 | |||
| BC064105.1 | - | 2 | 72 | |||

2610203C22Rik
Genomic Map