siRNA Details
Gene Symbol: 1700057H15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040774.1 | 4 | 382 | |||
GenBank mRNA | AK132134.1 | - | 4 | 382 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713246.1 | - | 4 | 382 | Chr. 4 - 124449902 - 124485959 [-] on Build GRCm38 |
