siRNA Details
| Gene Symbol: 1700057H15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040774.1 | 4 | 310 | |||
| GenBank mRNA | AK018866.1 | - | 3 | 263 | ||
| AK132134.1 | - | 4 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700057H15Rik| Gene Symbol: 1700057H15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040774.1 | 4 | 310 | |||
| GenBank mRNA | AK018866.1 | - | 3 | 263 | ||
| AK132134.1 | - | 4 | 310 | |||