siRNA Details
| Gene Symbol: Gm12159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045100.1 | 1, 2 | 50 | |||
| GenBank mRNA | AK041275.1 | - | 1, 2 | 51 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12159| Gene Symbol: Gm12159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045100.1 | 1, 2 | 50 | |||
| GenBank mRNA | AK041275.1 | - | 1, 2 | 51 | ||