siRNA Details
| Gene Symbol: 1810062O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033571.1 | 6 | 538 | |||
| GenBank mRNA | AK007938.1 | - | 6 | 540 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810062O18Rik| Gene Symbol: 1810062O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033571.1 | 6 | 538 | |||
| GenBank mRNA | AK007938.1 | - | 6 | 540 | ||