siRNA Details
| Gene Symbol: 2810471M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045906.1 | 1 | 515 | |||
| GenBank mRNA | AK013397.1 | - | 1 | 516 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810471M01Rik| Gene Symbol: 2810471M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045906.1 | 1 | 515 | |||
| GenBank mRNA | AK013397.1 | - | 1 | 516 | ||