siRNA Details
| Gene Symbol: Lyzl4os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040740.1 | 3 | 351 | |||
| GenBank mRNA | AK016372.1 | - | 3 | 374 | ||
| AK016399.1 | - | 3 | 352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyzl4os| Gene Symbol: Lyzl4os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040740.1 | 3 | 351 | |||
| GenBank mRNA | AK016372.1 | - | 3 | 374 | ||
| AK016399.1 | - | 3 | 352 | |||