siRNA Details
| Gene Symbol: Lyzl4os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040740.1 | 3, 4 | 442 | |||
| GenBank mRNA | AK016399.1 | - | 3, 4 | 443 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyzl4os| Gene Symbol: Lyzl4os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040740.1 | 3, 4 | 442 | |||
| GenBank mRNA | AK016399.1 | - | 3, 4 | 443 | ||