siRNA Details
| Gene Symbol: Gm15471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040412.1 | 4 | 314 | |||
| GenBank mRNA | AK052792.1 | - | 3 | 920 | ||
| BX516086.1 | - | 4 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15471| Gene Symbol: Gm15471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040412.1 | 4 | 314 | |||
| GenBank mRNA | AK052792.1 | - | 3 | 920 | ||
| BX516086.1 | - | 4 | 280 | |||