siRNA Details
| Gene Symbol: Gm15471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040412.1 | 3, 4 | 293 | |||
| GenBank mRNA | BX516086.1 | - | 3, 4 | 259 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15471| Gene Symbol: Gm15471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040412.1 | 3, 4 | 293 | |||
| GenBank mRNA | BX516086.1 | - | 3, 4 | 259 | ||