siRNA Details
| Gene Symbol: LOC102634401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110447.1 | 2 | 865 | |||
| GenBank mRNA | AK144266.1 | - | 2 | 866 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY755429.1 | - | 2 | 633 | Chr. 2 - 74752915 - 74753739 [+] on Build GRCm38 | |

LOC102634401
Genomic Map