siRNA Details
| Gene Symbol: Gm11240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046041.1 | 1 | 175 | |||
| GenBank mRNA | AK131821.1 | - | 1 | 177 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707593.1 | - | 1 | 173 | Chr. 4 - 73791246 - 73798352 [+] on Build GRCm38 | |

Gm11240
Genomic Map