siRNA Details
| Gene Symbol: Gm11240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046041.1 | 3 | 403 | |||
| NR_046042.1 | 3 | 332 | ||||
| GenBank mRNA | AK131821.1 | - | 3 | 405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11240| Gene Symbol: Gm11240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046041.1 | 3 | 403 | |||
| NR_046042.1 | 3 | 332 | ||||
| GenBank mRNA | AK131821.1 | - | 3 | 405 | ||