siRNA Details
| Gene Symbol: BB031773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028391.1 | 2 | 145 | |||
| GenBank mRNA | AK030945.1 | - | 2 | 147 | ||
| AK040278.1 | - | 2 | 144 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB031773| Gene Symbol: BB031773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028391.1 | 2 | 145 | |||
| GenBank mRNA | AK030945.1 | - | 2 | 147 | ||
| AK040278.1 | - | 2 | 144 | |||