siRNA Details
| Gene Symbol: Gm15834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053027.1 | - | 2, 3 | 1775 | ||
| AK144771.1 | - | 3, 4 | 282 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY734356.1 | - | 3, 4 | 282 | Chr. 1 - 58505150 - 58516120 [+] on Build GRCm38 | |

Gm15834
Genomic Map