siRNA Details
Gene Symbol: Saxo1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015482.1 | - | 1 | 85 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Saxo1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015482.1 | - | 1 | 85 |