siRNA Details
| Gene Symbol: 4933404K08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046038.1 | 4 | 667 | |||
| GenBank mRNA | AK016652.1 | - | 4 | 668 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716339.1 | - | 4 | 668 | Chr. 13 - 23278350 - 23281701 [+] on Build GRCm38 | |

4933404K08Rik
Genomic Map