siRNA Details
| Gene Symbol: 1110018N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003793.1 | - | 3 | 371 | ||
| AK004537.1 | - | 2 | 513 | |||
| AK044157.1 | - | 3 | 396 | |||
| AY672066.1 | - | 4 | 604 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110018N20Rik| Gene Symbol: 1110018N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003793.1 | - | 3 | 371 | ||
| AK004537.1 | - | 2 | 513 | |||
| AK044157.1 | - | 3 | 396 | |||
| AY672066.1 | - | 4 | 604 | |||