siRNA Details
| Gene Symbol: C130046K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102388.1 | 2 | 529 | |||
| NR_102389.1 | 2 | 529 | ||||
| NR_102396.1 | 2 | 529 | ||||
| GenBank mRNA | AK165205.1 | - | 2 | 530 | ||
| BE863793.1 | - | 2 | 409 | |||
| BY732345.1 | - | 2 | 530 | |||

C130046K22Rik
Genomic Map