siRNA Details
| Gene Symbol: 4930515L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045440.1 | 1 | 70 | |||
| GenBank mRNA | AK015799.1 | - | 1 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930515L19Rik| Gene Symbol: 4930515L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045440.1 | 1 | 70 | |||
| GenBank mRNA | AK015799.1 | - | 1 | 71 | ||