siRNA Details
| Gene Symbol: Gm12295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040280.1 | 1 | 133 | |||
| GenBank mRNA | AK042647.1 | - | 1 | 134 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12295| Gene Symbol: Gm12295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040280.1 | 1 | 133 | |||
| GenBank mRNA | AK042647.1 | - | 1 | 134 | ||