siRNA Details
| Gene Symbol: Gm10785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040389.1 | 6 | 897 | |||
| GenBank mRNA | AK038992.1 | - | 6 | 884 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10785| Gene Symbol: Gm10785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040389.1 | 6 | 897 | |||
| GenBank mRNA | AK038992.1 | - | 6 | 884 | ||