siRNA Details
| Gene Symbol: D830026I12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102304.1 | 3 | 628 | |||
| NR_102305.1 | 3 | 586 | ||||
| GenBank mRNA | AK081964.1 | - | 3 | 629 | ||
| CJ098230.1 | - | 3 | 587 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728579.1 | - | 3 | 629 | Chr. 6 - 17198107 - 17204751 [+] on Build GRCm38 | |

D830026I12Rik
Genomic Map