siRNA Details
Gene Symbol: Gm11423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158362.1 | - | 1 | 829 | ||
BM933646.1 | - | 2 | 126 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm11423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK158362.1 | - | 1 | 829 | ||
BM933646.1 | - | 2 | 126 |