siRNA Details
| Gene Symbol: 6030471H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020082.1 | - | 2 | 148 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6030471H07Rik| Gene Symbol: 6030471H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020082.1 | - | 2 | 148 | ||