siRNA Details
| Gene Symbol: Snhg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_003270.2 | 3 | 407 | |||
| GenBank mRNA | AJ006837.1 | - | LIT1313 U17HG |
3 | 353 | |
| AK076188.1 | - | 3 | 355 | |||
| BC100513.1 | - | 3 | 400 | |||
| CN834544.1 | - | 3 | 410 | |||

Snhg3
Genomic Map