siRNA Details
| Gene Symbol: Snhg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003270.2 | 1 | 20 | |||
| GenBank mRNA | BC100513.1 | - | 1 | 13 | ||
| CN834544.1 | - | 1 | 23 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snhg3| Gene Symbol: Snhg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003270.2 | 1 | 20 | |||
| GenBank mRNA | BC100513.1 | - | 1 | 13 | ||
| CN834544.1 | - | 1 | 23 | |||