siRNA Details
| Gene Symbol: 4930545L23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040517.1 | 6 | 2243 | |||
| GenBank mRNA | AK019742.1 | - | 6 | 2239 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB261654.1 | - | 1 | 28 | Chr. 2 - 135169573 - 135169859 [-] on Build GRCm38 | |

4930545L23Rik
Genomic Map