siRNA Details
| Gene Symbol: 4930545L23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040517.1 | 2 | 463 | |||
| GenBank mRNA | AK019742.1 | - | 2 | 459 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715640.1 | - | 2 | 458 | Chr. 2 - 135191459 - 135215616 [-] on Build GRCm38 | |

4930545L23Rik
Genomic Map