siRNA Details
| Gene Symbol: 8430437L04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040503.1 | 7 | 1297 | |||
| GenBank mRNA | AK018472.1 | - | 7 | 1297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

8430437L04Rik| Gene Symbol: 8430437L04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040503.1 | 7 | 1297 | |||
| GenBank mRNA | AK018472.1 | - | 7 | 1297 | ||