siRNA Details
| Gene Symbol: Gm15581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC067060.1 | - | 5 | 968 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15581| Gene Symbol: Gm15581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC067060.1 | - | 5 | 968 | ||