siRNA Details
| Gene Symbol: 4833418N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015506.2 | 2 | 824 | |||
| GenBank mRNA | AK014719.1 | - | 2 | 1049 | ||
| AK028291.1 | - | 1 | 2201 | |||
| AK031641.1 | - | 2 | 827 | |||
| AK082147.1 | - | 1 | 1112 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833418N02Rik| Gene Symbol: 4833418N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015506.2 | 2 | 824 | |||
| GenBank mRNA | AK014719.1 | - | 2 | 1049 | ||
| AK028291.1 | - | 1 | 2201 | |||
| AK031641.1 | - | 2 | 827 | |||
| AK082147.1 | - | 1 | 1112 | |||