siRNA Details
| Gene Symbol: 1700003M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040648.1 | 1 | 69 | |||
| GenBank mRNA | AA198428.1 | - | 1 | 80 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI390380.1 | - | 1 | 67 | Chr. 4 - 129960850 - 129963657 [+] on Build GRCm38 | |

1700003M07Rik
Genomic Map