siRNA Details
| Gene Symbol: Firre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030946.1 | - | 1 | 266 | ||
| BC066100.1 | - | 1 | 71 | |||
| BU704013.1 | - | 1 | 257 | |||
| KJ638684.1 | - | 1 | 30 | |||
| KJ638685.1 | - | 1 | 30 | |||
| KJ638686.1 | - | 1 | 225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Firre| Gene Symbol: Firre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030946.1 | - | 1 | 266 | ||
| BC066100.1 | - | 1 | 71 | |||
| BU704013.1 | - | 1 | 257 | |||
| KJ638684.1 | - | 1 | 30 | |||
| KJ638685.1 | - | 1 | 30 | |||
| KJ638686.1 | - | 1 | 225 | |||