siRNA Details
Gene Symbol: 1700019G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040255.1 | 3 | 433 | |||
GenBank mRNA | AK006121.1 | - | 3 | 400 | ||
AV266749.1 | - | 1 | 111 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700019G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040255.1 | 3 | 433 | |||
GenBank mRNA | AK006121.1 | - | 3 | 400 | ||
AV266749.1 | - | 1 | 111 |