siRNA Details
| Gene Symbol: 6430710C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102348.1 | 6 | 821 | |||
| GenBank mRNA | AK018302.1 | - | 6 | 776 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719186.1 | - | 6 | 777 | Chr. 2 - 72746516 - 72813672 [-] on Build GRCm38 | |

6430710C18Rik
Genomic Map