siRNA Details
Gene Symbol: 1700047A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110583.1 | 3 | 205 | |||
GenBank mRNA | AK006702.1 | - | 3 | 206 | ||
AK019763.1 | - | 3 | 199 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700047A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110583.1 | 3 | 205 | |||
GenBank mRNA | AK006702.1 | - | 3 | 206 | ||
AK019763.1 | - | 3 | 199 |