siRNA Details
Gene Symbol: Sorbs2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045739.1 | 3 | 1057 | |||
GenBank mRNA | AK084672.1 | - | 3 | 1058 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sorbs2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045739.1 | 3 | 1057 | |||
GenBank mRNA | AK084672.1 | - | 3 | 1058 |